missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzylpenicillin Procaine, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Benzylpenicillin Procaine
Specifications
Specifications
| Chemical Name or Material | Procaine benzylpenicillin hydrate |
| CAS | 6130-64-9 |
| Analyte or Component Names | Procaine benzylpenicillin hydrate |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C16 H18 N2 O4 S . C13 H20 N2 O2 . H2 O |
| Certifications/Compliance | ISO 17034 |
| Synonym | 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3,3-dimethyl-7-oxo-6-[(phenylacetyl)amino]- (2S,5R,6R)-, compd. with 2-(diethylamino)ethyl 4-aminobenzoate, hydrate (1:1:1), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3,3-dimethyl-7-oxo-6-(2-phenylacetamido)-, compd. with 2-(diethylamino)ethyl p-aminobenzoate (1:1), monohydrate (8CI), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3,3-dimethyl-7-oxo-6-[(phenylacetyl)amino]- (2S,5R,6R)-, compd. with 2-(diethylamino)ethyl 4-aminobenzoate (1:1), monohydrate (9CI), Benzoic acid, 4-amino-, 2-(diethylamino)ethyl ester, mono[(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate], monohydrate (9CI), Benzoic acid, p-amino-, 2-(diethylamino)ethyl ester, mono[3,3-dimethyl-7-oxo-6-(2-phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate], monohydrate (8CI), Benzylpenicillin procaine monohydrate, Procaine penicillin G hydrate, Procaine penicillin G monohydrate, 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3,3-dimethyl-7-oxo-6-[(phenylacetyl)amino]- [2S-(2alpha,5alpha,6beta)]-, compd. with 2-(diethylamino)ethyl 4-aminobenzoate (1:1), monohydrate, Benzoic acid, 4-amino-, 2-(diethylamino)ethyl ester, mono[[2S-(2alpha,5alpha,6beta)]-3,3-dimethyl-7-oxo-6-[(phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate], monohydrate |
| InChI Formula | InChI=1S/C16H18N2O4S.C13H20N2O2.H2O/c1-16(2)12(15(21)22)18-13(20)11(14(18)23-16)17-10(19)8-9-6-4-3-5-7-9;1-3-15(4-2)9-10-17-13(16)11-5-7-12(14)8-6-11;/h3-7,11-12,14H,8H2,1-2H3,(H,17,19)(H,21,22);5-8H,3-4,9-10,14H2,1-2H3;1H2/t11-,12+,14-;;/m1../s1 |
| SMILES | O.CCN(CC)CCOC(=O)c1ccc(N)cc1.CC2(C)S[C@@H]3[C@H](NC(=O)Cc4ccccc4)C(=O)N3[C@H]2C(=O)O |
| IUPAC Name | 2-(diethylamino)ethyl 4-aminobenzoate;(2S,5R,6R)-3,3-dimethyl-7-oxo-6-[(2-phenylacetyl)amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;hydrate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction