Learn More
Bromfenac Methyl Ester, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | Bromfenac Methyl Ester |
| CAS | 2445168-29-4 |
| Molecular Formula | C16 H14 Br N O3 |
| Quantity | 250 mg |
| Synonym | Bromfenac Methyl Ester, Benzeneacetic acid, 2-amino-3-(4-bromobenzoyl)-, methyl ester (ACI), Methyl 2-amino-3-(4-bromobenzoyl)benzeneacetate (ACI) |
| InChI Formula | InChI=1S/C16H14BrNO3/c1-21-14(19)9-11-3-2-4-13(15(11)18)16(20)10-5-7-12(17)8-6-10/h2-8H,9,18H2,1H3 |
| SMILES | COC(=O)Cc1cccc(C(=O)c2ccc(Br)cc2)c1N |
| IUPAC Name | methyl 2-[2-amino-3-(4-bromobenzoyl)phenyl]acetate |
| Molecular Weight (g/mol) | 348.19 |
| Formula Weight | 347.0157 |