missing translation for 'onlineSavingsMsg'
Learn More

Galantamine Hydrobromide, Mikromol™

Product Code. 18091900
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 18091900

Brand: Mikromol MM1256.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Galantamine Hydrobromide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Galantamine Hydrobromide
CAS 4-4-1953
Analyte or Component Names Galantamine Hydrobromide
Shipping Condition Room Temperature
Molecular Formula C17 H21 N O3 . Br H
Certifications/Compliance ISO 17034
Synonym 6H-Benzofuro[3a,3,2-ef][2]benzazepin-6-ol, 4a,5,9,10,11,12-hexahydro-3-methoxy-11-methyl-, hydrobromide (1:1), (4aS,6R,8aS)-, 6H-Benzofuro[3a,3,2-ef][2]benzazepin-6-ol, 4a,5,9,10,11,12-hexahydro-3-methoxy-11-methyl-, hydrobromide, (4aS,6R,8aS)- (9CI), Galanthamine, hydrobromide (6CI,8CI), (-)-Galantamine hydrobromide, (-)-Galanthamine hydrobromide, Galantamine hydrobromide, Galanthamine hydrogen bromide, Jilkon hydrobromide, Lycoremine hydrobromide, Nivalin, Nivaline, Nivaline (pharmaceutical), Razadyne, Reminyl, Tamilin, (4aS,6R,8aS)-3-Methoxy-11-methyl-5,6,9,10,11,12-hexahydro-4aH-[1]benzofuro[3a,3,2-ef][2]benzazepin-6-ol hydrobromide
InChI Formula InChI=1S/C17H21NO3.BrH/c1-18-8-7-17-6-5-12(19)9-14(17)21-16-13(20-2)4-3-11(10-18)15(16)17;/h3-6,12,14,19H,7-10H2,1-2H3;1H/t12-,14-,17-;/m0./s1
SMILES Br.COc1ccc2CN(C)CC[C@@]34C=C[C@H](O)C[C@@H]3Oc1c24
IUPAC Name (4aS,6R,8aS)-4a,5,9,10,11,12-Hexahydro-3-methoxy-11-methyl-6H-benzofuro[3a,3,2-ef][2]benzazepin-6-ol hydrobromide
Molecular Weight (g/mol) 368.27
Quantity 250 mg
Formula Weight 367.0783 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.