missing translation for 'onlineSavingsMsg'
Learn More

Ketoprofen Lysinate, Mikromol™

Product Code. 17979719
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 17979719

Brand: Mikromol MM3602.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Ketoprofen Lysinate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ketoprofen Lysinate
CAS 57469-78-0
Analyte or Component Names Ketoprofen Lysinate
Shipping Condition Room Temperature
Molecular Formula C16 H14 O3 . C6 H14 N2 O2
Certifications/Compliance ISO 17034
Synonym Ketoprofen Lysinate, 3-Benzoyl-alpha-methylbenzeneacetate L-Lysine (1:1), L-Lysine, mono(3-benzoyl-alpha-methylbenzeneacetate) (9CI), Benzeneacetic acid, 3-benzoyl-alpha-methyl-, compd. with L-lysine (1:1) (9CI), Artrosilene, Ketoprofen L-lysinate, Ketoprofen lysinate, Ketoprofen lysine salt, L-Lysine, 3-benzoyl-alpha-methylbenzeneacetate, Lysine m-benzoylhydratropate, OKI spray, OKi
InChI Formula InChI=1S/C16H14O3.C6H14N2O2/c1-11(16(18)19)13-8-5-9-14(10-13)15(17)12-6-3-2-4-7-12;7-4-2-1-3-5(8)6(9)10/h2-11H,1H3,(H,18,19);5H,1-4,7-8H2,(H,9,10)/t;5-/m.0/s1
SMILES CC(C(=O)O)c1cccc(c1)C(=O)c2ccccc2.NCCCC[C@H](N)C(=O)O
IUPAC Name 2-(3-benzoylphenyl)propanoic acid;(2S)-2,6-diaminohexanoic acid
Molecular Weight (g/mol) 400.47
Quantity 250 mg
Formula Weight 400.1998 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.