missing translation for 'onlineSavingsMsg'
Learn More

Prednisolone Acetate, Mikromol™

Product Code. 17951376
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 17951376

Brand: Mikromol MM0194.000250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Prednisolone Acetate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Prednisolone Acetate
CAS 52-21-1
Analyte or Component Names Prednisolone Acetate
Shipping Condition Room Temperature
Molecular Formula C23 H30 O6
Certifications/Compliance ISO 17034
Synonym Hydrocortisone Acetate Imp. C (EP), Methylprednisolone Acetate Imp. E (EP), Prednisolone Imp. C (EP), Prednisolone Acetate, 11beta,17-Dihydroxy-3,20-dioxo-pregna-1,4-dien-21-yl Acetate, Hydrocortisone Acetate Imp. C (Pharmeuropa), (11beta)-21-(Acetyloxy)-11,17-dihydroxypregna-1,4-diene-3,20-dione, 11beta,17alpha,21-Trihydroxypregna-1,4-diene-3,20-dione 21-acetate, 21-(Acetoxy)-11beta,17alpha-dihydroxypregna-1,4-diene-3,20-dione, 21-Acetoxy-11beta,17-dihydroxypregna-1,4-diene-3,20-dione, Ak-Tate, Cormalone, Cortipred, Deltilen, Econopred, Falcon, Falcon (steroid), Hydroprednisone acetate, Inflanefran, Inflanefran Forte, Meticortelone acetate, Meticotelone acetate, NSC 10966, Nisolone, Pred Mild, Pred-Forte, Predalone 50, Prediacortin, Predicort, Prednelan N, Prednidoren, Prednisolone 21-acetate, Prednisolone acetate, Prenema, Supercortyl, Hydrocortisone Acetate Impurity C, Methylprednisolone Acetate Impurity E, Prednisolone Impurity C, Prednisone Impurity L
InChI Formula InChI=1S/C23H30O6/c1-13(24)29-12-19(27)23(28)9-7-17-16-5-4-14-10-15(25)6-8-21(14,2)20(16)18(26)11-22(17,23)3/h6,8,10,16-18,20,26,28H,4-5,7,9,11-12H2,1-3H3/t16-,17-,18-,20+,21-,22-,23-/m0/s1
SMILES CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3[C@@H](O)C[C@]12C
IUPAC Name [2-[(8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
Molecular Weight (g/mol) 402.48
Quantity 250 mg
Formula Weight 402.2042 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.