missing translation for 'onlineSavingsMsg'
Learn More

Propionylpromazine Hydrochloride, TRC

Product Code. 17964045
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17964045 250 mg 250mg
17993475 100 mg 100mg
17924125 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17964045 Supplier Toronto Research Chemicals Supplier No. TRCP827500250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-67-6
Molecular Formula C20 H24 N2 O S . Cl H
Molecular Weight (g/mol) 376.94
Synonym Propionylpromazine Hydrochloride, 1-Propanone, 1-[10-[3-(dimethylamino)propyl]-10H-phenothiazin-2-yl]-, monohydrochloride (9CI), 1-Propanone, 1-[10-[3-(dimethylamino)propyl]phenothiazin-2-yl]-, hydrochloride (7CI), 1-Propanone, 1-[10-[3-(dimethylamino)propyl]phenothiazin-2-yl]-, monohydrochloride (8CI), 2-Propionyl-10-(3-dimethylaminopropyl)phenothiazine hydrochloride, Propiopromazine hydrochloride, Tranvet, 1-Propanone, 1-[10-[3-(dimethylamino)propyl]-10H-phenothiazin-2-yl]-, hydrochloride (1:1)
IUPAC Name 1-[10-[3-(dimethylamino)propyl]phenothiazin-2-yl]propan-1-one;hydrochloride
SMILES Cl.CCC(=O)c1ccc2Sc3ccccc3N(CCCN(C)C)c2c1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C20H24N2OS.ClH/c1-4-18(23)15-10-11-20-17(14-15)22(13-7-12-21(2)3)16-8-5-6-9-19(16)24-20;/h5-6,8-11,14H,4,7,12-13H2,1-3H3;1H
Formula Weight 376.1376
Chemical Name or Material Propionylpromazine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.