missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rivastigmine Hydrogen Tartrate, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Rivastigmine Hydrogen Tartrate
Specifications
Specifications
| Chemical Name or Material | Rivastigmine hydrogen tartrate |
| CAS | 129101-54-8 |
| Analyte or Component Names | Rivastigmine hydrogen tartrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H22 N2 O2 . C4 H6 O6 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Rivastigmine Tartrate, Rivastigmine hydrogen tartrate, 3-[(1S)-1-(Dimethylamino)ethyl]phenyl ethyl(methyl)carbamate hydrogen (2R,3R)-2,3-dihydroxybutanedioate, Carbamic acid, N-ethyl-N-methyl-, 3-[(1S)-1-(dimethylamino)ethyl]phenyl ester, (2R,3R)-2,3-dihydroxybutanedioate (1:1)Carbamic acid, ethylmethyl-, 3-[(1S)-1-(dimethylamino)ethyl]phenyl ester, (2R,3R)-2,3-dihydroxybutanedioate (1:1) (9CI), Carbamic acid, ethylmethyl-, 3-[1-(dimethylamino)ethyl]phenyl ester, (S)-, [R-(R*,R*)]-2,3-dihydroxybutanedioate (1:1), ENA 713, Rivastigmine hydrogentartrate, Rivastigmine tartrate, SDZ-ENA 713, rivastigmine bitartrate |
| InChI Formula | InChI=1S/C14H22N2O2.C4H6O6/c1-6-16(5)14(17)18-13-9-7-8-12(10-13)11(2)15(3)4;5-1(3(7)8)2(6)4(9)10/h7-11H,6H2,1-5H3;1-2,5-6H,(H,7,8)(H,9,10)/t11-;1-,2-/m01/s1 |
| SMILES | CCN(C)C(=O)Oc1cccc(c1)[C@H](C)N(C)C.O[C@H]([C@@H](O)C(=O)O)C(=O)O |
| IUPAC Name | (2R,3R)-2,3-dihydroxybutanedioic acid;[3-[(1S)-1-(dimethylamino)ethyl]phenyl] N-ethyl-N-methylcarbamate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction