missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Testosterone Decanoate, Mikromol™
Description
Testosterone Decanoate
Specifications
Specifications
| Chemical Name or Material | Testosterone decanoate |
| CAS | 5721-91-5 |
| Analyte or Component Names | Testosterone decanoate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C29 H46 O3 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Testosterone Decanoate, Decanoic acid, ester with testosterone, NSC 26642, Testosterone 17-caprinate, Testosterone 17-decanoate, Testosterone caprate, Testosterone caprinate, (17beta)-17-[(1-Oxodecyl)oxy]androst-4-en-3-one, 17beta-Hydroxyandrost-4-en-3-one decanoate |
| InChI Formula | InChI=1S/C29H46O3/c1-4-5-6-7-8-9-10-11-27(31)32-26-15-14-24-23-13-12-21-20-22(30)16-18-28(21,2)25(23)17-19-29(24,26)3/h20,23-26H,4-19H2,1-3H3/t23-,24-,25-,26-,28-,29-/m0/s1 |
| SMILES | CCCCCCCCCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C |
| IUPAC Name | [(8R,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] decanoate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?