missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vanillylmethanol, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | Vanillylmethanol |
| CAS | 2380-78-1 |
| Molecular Formula | C9 H12 O3 |
| Quantity | 250 mg |
| Synonym | 2-(3-Methoxy-4-hydroxyphenyl)ethanol, 2-(4-Hydroxy-3-methoxyphenyl)ethanol, 3-Methoxy-4-hydroxyphenethanol, 3-Methoxy-4-hydroxyphenethyl Alcohol, 4-(2-Hydroxyethyl)-2-methoxyphenol, 4-(2-Hydroxyethyl)guaiacol, 4-Hydroxy-3-methoxyphenethyl Alcohol, 4-Hydroxy-3-methoxyphenylethanol, Ba 2772, Homovanillic Alcohol, Homovanillyl Alcohol |
| InChI Formula | InChI=1S/C9H12O3/c1-12-9-6-7(4-5-10)2-3-8(9)11/h2-3,6,10-11H,4-5H2,1H3 |
| SMILES | COc1cc(CCO)ccc1O |
| IUPAC Name | 4-(2-hydroxyethyl)-2-methoxyphenol |
| Molecular Weight (g/mol) | 168.19 |
| Formula Weight | 168.079 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction