Learn More
Reagecon 250ML Ion Chromatography Multi Element (5) CationStandard Ca, K, Mg, Na, NH4 @0.5ppm each in Water
Brand: Reagecon ICC5MIX3B
250ML Ion Chromatography Multi Element (5) CationStandard Ca, K, Mg, Na, NH4 @0.5ppm each in Water