missing translation for 'onlineSavingsMsg'
Learn More

Uranium Standard: U @ 1000 μg/mL in 5% HNO3, VHG™

Product Code. 17937679
Click to view available options
Quantity:
50 mL
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
50mL
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 17937679

Brand: VHG Labs VHGPUN250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Uranium Standard: U @ 1000 μg/mL in 5% HNO3
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Uranium
CAS 7440-61-1
Methods ICP/ICP-MS
Analyte or Component Names Uranium
Shipping Condition Room Temperature
Molecular Formula U
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym U
InChI Formula InChI=1S/U
SMILES [U]
IUPAC Name uranium
Solvent or Matrix Dilute HNO3
Molecular Weight (g/mol) 238.029
Quantity 250 mL
Formula Weight 238.051 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.