Learn More
n-Propyl Alcohol-OD, CDN
Product Code. 30316120
Click to view available options
Quantity:
25 g
Unit Size:
25g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30316120
Brand: CDN CDND008125G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-Propanol D (OD)
CAS 4712-36-1
Molecular Formula C3 D H7 O
Quantity 25 g
Synonym 1-Propanol D (OD), Propyl alcohol-d (6 CI,7 CI,8 CI), 1-Propanol-O-d, Propanol-d, n-Propanol-d, 1-Propanol-d (9 CI)
InChI Formula InChI=1 S/C3H8O/c1-2-3-4/h4H,2-3H2,1H3/i4D
SMILES [2 H]OCCC
IUPAC Name 1-deuteriooxypropane
Molecular Weight (g/mol) 61.101
Formula Weight 61.064 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity