missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cimetidine EP Impurity H, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
Cimetidine EP Impurity H
Specifications
Specifications
| Chemical Name or Material | 1,1'-(Disulfanediyldiethylene)bis(2-cyano-3-methylguanidine) |
| CAS | 74886-59-2 |
| Analyte or Component Names | 1,1'-(Disulfanediyldiethylene)bis(2-cyano-3-methylguanidine) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H18 N8 S2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 1,1'-(Disulphanediyldiethylene)bis(2-cyano-3-methylguanidine), Cimetidine Hydrochloride Imp. H (EP), Cimetidine Imp. H (EP), 1,1'-(Disulphanediyldiethylene)bis(2-cyano-3-methylguanidine), Cimetidine Hydrochloride Impurity H, Cimetidine Impurity H, Cimetidine Impurity H |
| InChI Formula | InChI=1S/C10H18N8S2/c1-13-9(17-7-11)15-3-5-19-20-6-4-16-10(14-2)18-8-12/h3-6H2,1-2H3,(H2,13,15,17)(H2,14,16,18) |
| SMILES | CNC(=NC#N)NCCSSCCNC(=NC#N)NC |
| IUPAC Name | 2-cyano-1-[2-[2-[(N'-cyano-N-methylcarbamimidoyl)amino]ethyldisulfanyl]ethyl]-3-methylguanidine |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction