missing translation for 'onlineSavingsMsg'
Learn More

[1-[[[(1R)-1-[3-[(E)-2-(7-Chloroquinolin-2-yl)ethenyl]phenyl]-3-[2-(1-methylethenyl)phenyl]propyl]sulfanyl]methyl]cyclopropyl]acetic Acid, Mikromol™

Product Code. 30051803
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 30051803

Brand: Mikromol MM0698.020025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

[1-[[[(1R)-1-[3-[(E)-2-(7-Chloroquinolin-2-yl)ethenyl]phenyl]-3-[2-(1-methylethenyl)phenyl]propyl]sulfanyl]methyl]cyclopropyl]acetic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Montelukast Styrene
CAS 918972-54-0
Analyte or Component Names Montelukast Styrene
Shipping Condition Room Temperature
Molecular Formula C35 H34 Cl N O2 S
Certifications/Compliance ISO 9001
Synonym Montelukast Sodium Imp. B (EP), Montelukast Imp. B (EP), 1-[[[(1R)-1-[3-[(1E)-2-(7-Chloro-2-quinolinyl)ethenyl]phenyl]-3-[2-(1-methylethenyl)phenyl]propyl]thio]methyl]-cyclopropaneacetic Acid, Montelukast Styrene, Montekulast Sodium Imp. B (EP), Montelukast Sodium Impurity B, Montelukast Impurity B
InChI Formula InChI=1S/C35H34ClNO2S/c1-24(2)31-9-4-3-7-26(31)13-17-33(40-23-35(18-19-35)22-34(38)39)28-8-5-6-25(20-28)10-15-30-16-12-27-11-14-29(36)21-32(27)37-30/h3-12,14-16,20-21,33H,1,13,17-19,22-23H2,2H3,(H,38,39)/b15-10+/t33-/m1/s1
SMILES CC(=C)c1ccccc1CC[C@@H](SCC2(CC(=O)O)CC2)c3cccc(\C=C\c4ccc5ccc(Cl)cc5n4)c3
IUPAC Name 2-[1-[[(1R)-1-[3-[(E)-2-(7-chloroquinolin-2-yl)ethenyl]phenyl]-3-(2-prop-1-en-2-ylphenyl)propyl]sulfanylmethyl]cyclopropyl]acetic acid
Molecular Weight (g/mol) 568.17
Quantity 25 mg
Formula Weight 567.1999 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.