missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,3,4-Tetrahydro-4,6-dihydroxy-2-methyl-isoquinoline Hydrochloride, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
1,2,3,4-Tetrahydro-4,6-dihydroxy-2-methyl-isoquinoline Hydrochloride
Specifications
Specifications
| Chemical Name or Material | 1,2,3,4-Tetrahydro-4,6-dihydroxy-2-methyl-isoquinoline Hydrochloride |
| CAS | 57196-61-9 |
| Analyte or Component Names | 1,2,3,4-Tetrahydro-4,6-dihydroxy-2-methyl-isoquinoline Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H13 N O2 . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | 1,2,3,4-Tetrahydro-4,6-dihydroxy-2-methyl-isoquinoline Hydrochloride |
| InChI Formula | InChI=1S/C10H13NO2.ClH/c1-11-5-7-2-3-8(12)4-9(7)10(13)6-11;/h2-4,10,12-13H,5-6H2,1H3;1H |
| SMILES | Cl.CN1CC(O)c2cc(O)ccc2C1 |
| IUPAC Name | 2-methyl-3,4-dihydro-1H-isoquinoline-4,6-diol;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction