missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(4-Amino-6,7-dimethoxyquinazolin-2-yl)-4-formylpiperazine Hydrochloride, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
1-(4-Amino-6,7-dimethoxyquinazolin-2-yl)-4-formylpiperazine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | 1-(4-Amino-6,7-dimethoxyquinazolin-2-yl)-4-formylpiperazine Hydrochloride |
| CAS | 84050-21-5 |
| Analyte or Component Names | 1-(4-Amino-6,7-dimethoxyquinazolin-2-yl)-4-formylpiperazine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H19 N5 O3 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Terazosin Hydrochloride Dihydrate Imp. D (EP), Terazosin Imp. D (EP), 1-(4-Amino-6,7-dimethoxyquinazolin-2-yl)-4-formylpiperazine Hydrochloride, Terazosin Hydrochloride Dihydrate Imp. D (EP) as Hydrochloride, Terazosin Hydrochloride Dihydrate Impurity D as Hydrochloride, Terazosin Impurity D as Hydrochloride |
| InChI Formula | InChI=1S/C15H19N5O3.ClH/c1-22-12-7-10-11(8-13(12)23-2)17-15(18-14(10)16)20-5-3-19(9-21)4-6-20;/h7-9H,3-6H2,1-2H3,(H2,16,17,18);1H |
| SMILES | Cl.COc1cc2nc(nc(N)c2cc1OC)N3CCN(CC3)C=O |
| IUPAC Name | 4-(4-amino-6,7-dimethoxyquinazolin-2-yl)piperazine-1-carbaldehyde;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction