missing translation for 'onlineSavingsMsg'
Learn More

Azacitidine USP Related Compound B, Mikromol™

Product Code. 30049541
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 30049541

Brand: Mikromol MM1259.030025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Azacitidine USP Related Compound B
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-O-Acetyl-2,3,5-tri-O-benzoyl-beta-D-ribofuranose
CAS 6974-32-9
Analyte or Component Names 1-O-Acetyl-2,3,5-tri-O-benzoyl-beta-D-ribofuranose
Shipping Condition Room Temperature
Molecular Formula C28 H24 O9
Certifications/Compliance ISO 17025
Synonym 1-O-Acetyl-2,3,5-tri-O-benzoyl-β-D-ribofuranose, ABR, NSC 23349, beta-D-Ribofuranose, 1-acetate 2,3,5-tribenzoate, Ribofuranose, 1-acetate 2,3,5-tribenzoate, beta-D- (6CI,8CI), 1-0-Acetyl-2,3,5-tri-O-benzoyl-beta-D-ribofuranose, 1-O-Acetyl-2,3,5-tri-O-benzoyl-beta-D-ribofuranose, 1-O-Acetyl-2,3,5-tri-O-benzoyl-beta-D-ribose, 1-O-Acetyltri-O-benzoyl-beta-D-ribofuranose, 2,3,5-Tri-O-benzoyl-beta-D-ribofuranosyl acetate, Azacitidine Related Compound B
InChI Formula InChI=1S/C28H24O9/c1-18(29)34-28-24(37-27(32)21-15-9-4-10-16-21)23(36-26(31)20-13-7-3-8-14-20)22(35-28)17-33-25(30)19-11-5-2-6-12-19/h2-16,22-24,28H,17H2,1H3/t22-,23-,24-,28-/m1/s1
SMILES CC(=O)O[C@@H]1O[C@H](COC(=O)c2ccccc2)[C@@H](OC(=O)c3ccccc3)[C@H]1OC(=O)c4ccccc4
IUPAC Name [(2R,3R,4R,5S)-5-acetyloxy-3,4-dibenzoyloxyoxolan-2-yl]methyl benzoate
Molecular Weight (g/mol) 504.48
Quantity 25 mg
Formula Weight 504.142 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.