missing translation for 'onlineSavingsMsg'
Learn More
Learn More
16alpha,17-[(1RS)-Butylidenebis(oxy)]-11beta,21-dihydroxypregna-1,4-diene-3,20-dione 21-Acetate, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
16alpha,17-[(1RS)-Butylidenebis(oxy)]-11beta,21-dihydroxypregna-1,4-diene-3,20-dione 21-Acetate
Specifications
Specifications
| Chemical Name or Material | 16alpha,17-[(1RS)-Butylidenebis(oxy)]-11beta,21-dihydroxypregna-1,4-diene-3,20-dione 21-Acetate |
| CAS | 51333-05-2 |
| Analyte or Component Names | 16alpha,17-[(1RS)-Butylidenebis(oxy)]-11beta,21-dihydroxypregna-1,4-diene-3,20-dione 21-Acetate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C27 H36 O7 |
| Certifications/Compliance | ISO 9001 |
| Synonym | (11β,16α)-21-(Acetyloxy)-16,17-[butylidenebis(oxy)]-11-hydroxy-pregna-1,4-diene-3,20-dione, 21-(Acetoxy)-16α,17α-(butylidenedioxy)-11β-hydroxypregna-1,4-diene-3,20-dione, 16α,17-[(1RS)-Butylidenebis(oxy)]-11β,21-dihydroxypregna-1,4-diene-3,20-dione 21-Acetate, Budesonide Imp. K (EP) |
| InChI Formula | InChI=1S/C27H36O7/c1-5-6-23-33-22-12-19-18-8-7-16-11-17(29)9-10-25(16,3)24(18)20(30)13-26(19,4)27(22,34-23)21(31)14-32-15(2)28/h9-11,18-20,22-24,30H,5-8,12-14H2,1-4H3/t18-,19-,20-,22+,23?,24+,25-,26-,27+/m0/s1 |
| SMILES | CCCC1O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@H]4[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)COC(=O)C |
| Molecular Weight (g/mol) | 472.57 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction