missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1H-1,2,4-Triazole-3-carboxylic Acid, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
1H-1,2,4-Triazole-3-carboxylic Acid
Specifications
Specifications
| Chemical Name or Material | 1H-1,2,4-Triazole-3-carboxylic Acid |
| CAS | 4928-87-4 |
| Analyte or Component Names | 1H-1,2,4-Triazole-3-carboxylic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C3 H3 N3 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Ribavirin Imp. C (EP), 1H-1,2,4-Triazole-3-carboxylic Acid, Ribavirin Impurity C |
| InChI Formula | InChI=1S/C3H3N3O2/c7-3(8)2-4-1-5-6-2/h1H,(H,7,8)(H,4,5,6) |
| SMILES | OC(=O)c1nc[nH]n1 |
| IUPAC Name | 1H-1,2,4-triazole-3-carboxylic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction