missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sibutramine EP Impurity I, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Sibutramine EP Impurity I
Specifications
Specifications
| Chemical Name or Material | (1RS)-1-(1-(4-Chlorophenyl)cyclobutyl)-3-methylbutan-1-ol |
| Analyte or Component Names | (1RS)-1-(1-(4-Chlorophenyl)cyclobutyl)-3-methylbutan-1-ol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H21 Cl O |
| Certifications/Compliance | ISO 17025 |
| Synonym | (1RS)-1-[1-(4-Chlorophenyl)cyclobutyl]-3-methylbutan-1-ol, Sibutramine Hydrochloride Monohydrate Imp. I (Pharmeuropa), Sibutramine Hydrochloride Monohydrate Impurity I, Sibutramine Impurity I |
| InChI Formula | InChI=1S/C15H21ClO/c1-11(2)10-14(17)15(8-3-9-15)12-4-6-13(16)7-5-12/h4-7,11,14,17H,3,8-10H2,1-2H3 |
| SMILES | CC(C)CC(O)C1(CCC1)c2ccc(Cl)cc2 |
| IUPAC Name | 1-[1-(4-chlorophenyl)cyclobutyl]-3-methylbutan-1-ol |
| Molecular Weight (g/mol) | 252.78 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction