missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1RS,2RS)-2-[(Dimethylamino)methyl]-1-(3-hydroxyphenyl)cyclohexanol Hydrochloride, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
(1RS,2RS)-2-[(Dimethylamino)methyl]-1-(3-hydroxyphenyl)cyclohexanol Hydrochloride
Specifications
Specifications
| Chemical Name or Material | O-Desmethyltramadol Hydrochloride |
| CAS | 185453-02-5 |
| Analyte or Component Names | O-Desmethyltramadol Hydrochloride |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C15 H23 N O2 . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | (1RS,2RS)-2-[(Dimethylamino)methyl]-1-(3-hydroxyphenyl)cyclohexanol Hydrochloride, Tramadol Hydrochloride Imp. D (EP) as Hydrochloride, O-Desmethyltramadol Hydrochloride, Tramadol Hydrochloride Imp. D (EP), Tramadol Imp. D (EP), Phenol, 3-[(1R,2R)-2-[(dimethylamino)methyl]-1-hydroxycyclohexyl]-, hydrochloride (1:1), rel-, Phenol, 3-[(1R,2R)-2-[(dimethylamino)methyl]-1-hydroxycyclohexyl]-, hydrochloride, rel- (9CI), Phenol, 3-[2-[(dimethylamino)methyl]-1-hydroxycyclohexyl]-, hydrochloride, cis-, O-Demethyltramadol hydrochloride, Tramadol Hydrochloride Impurity D as Hydrochloride, Tramadol Impurity D as Hydrochloride |
| InChI Formula | InChI=1S/C15H23NO2.ClH/c1-16(2)11-13-6-3-4-9-15(13,18)12-7-5-8-14(17)10-12;/h5,7-8,10,13,17-18H,3-4,6,9,11H2,1-2H3;1H/t13-,15+;/m1./s1 |
| SMILES | Cl.CN(C)C[C@H]1CCCC[C@]1(O)c2cccc(O)c2 |
| IUPAC Name | 3-[(1R,2R)-2-[(dimethylamino)methyl]-1-hydroxycyclohexyl]phenol;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction