missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-[4-(1-Hydroxy-2-methylpropyl)phenyl]propanoic Acid (1-Hydroxyibuprofen), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
2-[4-(1-Hydroxy-2-methylpropyl)phenyl]propanoic Acid (1-Hydroxyibuprofen)
Specifications
Specifications
| Chemical Name or Material | 2-[4-(1-Hydroxy-2-methylpropyl)phenyl]propanoic Acid (1-Hydroxyibuprofen) |
| CAS | 53949-53-4 |
| Analyte or Component Names | 2-[4-(1-Hydroxy-2-methylpropyl)phenyl]propanoic Acid (1-Hydroxyibuprofen) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H18 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 2-[4-(1-Hydroxy-2-methylpropyl)phenyl]propanoic Acid, Ibuprofen Imp. L (EP), 1-Hydroxyibuprofen, 4-(1-Hydroxy-2-methylpropyl)-alpha-methylbenzeneacetic acid, 2-[4'-(1-Hydroxy-2-methylpropyl)phenyl]propionic acid, Ibuprofen Impurity L |
| InChI Formula | InChI=1S/C13H18O3/c1-8(2)12(14)11-6-4-10(5-7-11)9(3)13(15)16/h4-9,12,14H,1-3H3,(H,15,16) |
| SMILES | CC(C)C(O)c1ccc(cc1)C(C)C(=O)O |
| IUPAC Name | 2-[4-(1-hydroxy-2-methylpropyl)phenyl]propanoic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction