missing translation for 'onlineSavingsMsg'
Learn More

2-[[(4-Methoxy-3,5-dimethylpyridin-2-yl)-methyl]sulphinyl]-1H-benzimidazole, Mikromol™

Product Code. 30052147
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mg
100 mg
missing translation for 'unitSize'
100mg
25mg
This item is not returnable. View return policy

Product Code. 30052147

missing translation for 'mfr': Mikromol MM0095.230025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-[[(4-Methoxy-3,5-dimethylpyridin-2-yl)-methyl]sulphinyl]-1H-benzimidazole
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[((4-Methoxy-3,5-dimethylpyridin-2-yl)-methyl)sulphinyl]-1H-benzimidazole
CAS 73590-60-0
Analyte or Component Names 2-[((4-Methoxy-3,5-dimethylpyridin-2-yl)-methyl)sulphinyl]-1H-benzimidazole
Shipping Condition Ice pack (-20°C)
Molecular Formula C16 H17 N3 O2 S
Certifications/Compliance ISO 9001
Synonym 2-[[(4-Methoxy-3,5-dimethylpyridin-2-yl)-methyl]sulphinyl]-1H-benzimidazole
InChI Formula InChI=1S/C16H17N3O2S/c1-10-8-17-14(11(2)15(10)21-3)9-22(20)16-18-12-6-4-5-7-13(12)19-16/h4-8H,9H2,1-3H3,(H,18,19)
SMILES COc1c(C)cnc(CS(=O)c2nc3ccccc3[nH]2)c1C
IUPAC Name 2-[(4-methoxy-3,5-dimethylpyridin-2-yl)methylsulfinyl]-1H-benzimidazole
Molecular Weight (g/mol) 315.39
Quantity 25 mg
Formula Weight 315.1041 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.