Learn More
Bromazepam EP Impurity A, Mikromol™
Product Code. 30052687
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Product Code. 30052687
Brand: Mikromol MM0064.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Bromazepam EP Impurity A
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (2-Amino-5-bromophenyl)-(pyridin-2-yl)methanone
CAS 1563-56-0
Analyte or Component Names (2-Amino-5-bromophenyl)-(pyridin-2-yl)methanone
Shipping Condition Room Temperature
Molecular Formula C12 H9 Br N2 O
Certifications/Compliance ISO 17025
Synonym (2-Amino-5-bromophenyl)-(pyridin-2-yl)methanone, Bromazepam Imp. A (EP), Bromazepam Impurity A
InChI Formula InChI=1S/C12H9BrN2O/c13-8-4-5-10(14)9(7-8)12(16)11-3-1-2-6-15-11/h1-7H,14H2
SMILES Nc1ccc(Br)cc1C(=O)c2ccccn2
IUPAC Name (2-amino-5-bromophenyl)-pyridin-2-ylmethanone
Molecular Weight (g/mol) 277.12
Quantity 25 mg
Formula Weight 275.9898 g/mol