missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Cyclohexylcarbonyl)-2,3,6,7-tetrahydro-4H-pyrazino[2,1-a]isoquinolin-4-one, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
2-(Cyclohexylcarbonyl)-2,3,6,7-tetrahydro-4H-pyrazino[2,1-a]isoquinolin-4-one
Specifications
Specifications
| Chemical Name or Material | 2-(cyclohexylcarbonyl)-2,3,6,7-tetrahydro-4H-pyrazino [2,1-a]isoquinolin-4-one |
| CAS | 125273-86-1 |
| Analyte or Component Names | 2-(cyclohexylcarbonyl)-2,3,6,7-tetrahydro-4H-pyrazino [2,1-a]isoquinolin-4-one |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H22 N2 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Praziquantel Imp. B (EP), Praziquantel USP Related Compound B, Praziquantel USP RC B, 2-(Cyclohexylcarbonyl)-2,3,6,7-tetrahydro-4H-pyrazino[2,1-a]isoquinolin-4-one, Praziquantel RC B (USP), Praziquantel Related Compound B, Praziquantel Impurity B |
| InChI Formula | InChI=1S/C19H22N2O2/c22-18-13-20(19(23)15-7-2-1-3-8-15)12-17-16-9-5-4-6-14(16)10-11-21(17)18/h4-6,9,12,15H,1-3,7-8,10-11,13H2 |
| SMILES | O=C(C1CCCCC1)N2CC(=O)N3CCc4ccccc4C3=C2 |
| IUPAC Name | 2-(cyclohexanecarbonyl)-6,7-dihydro-3H-pyrazino[2,1-a]isoquinolin-4-one |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction