missing translation for 'onlineSavingsMsg'
Learn More

2-Deoxy-2,2-difluoro-D-erythro-pentofuranos-1-ulose-3,5-dibenzoate, Mikromol™

Product Code. 30052100
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 30052100

Brand: Mikromol MM1257.080025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-Deoxy-2,2-difluoro-D-erythro-pentofuranos-1-ulose-3,5-dibenzoate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Deoxy-2,2-difluoro-D-erythro-pentofuranos-1-ulose-3,5-dibe
CAS 122111-01-7
Analyte or Component Names 2-Deoxy-2,2-difluoro-D-erythro-pentofuranos-1-ulose-3,5-dibe
Shipping Condition Room Temperature
Molecular Formula C19 H14 F2 O6
Certifications/Compliance ISO 9001
InChI Formula InChI=1S/C19H14F2O6/c20-19(21)15(27-17(23)13-9-5-2-6-10-13)14(26-18(19)24)11-25-16(22)12-7-3-1-4-8-12/h1-10,14-15H,11H2/t14-,15+/m0/s1
SMILES FC1(F)[C@H](OC(=O)c2ccccc2)[C@H](COC(=O)c3ccccc3)OC1=O
IUPAC Name [(2S,3R)-3-benzoyloxy-4,4-difluoro-5-oxooxolan-2-yl]methyl benzoate
Molecular Weight (g/mol) 376.31
Quantity 25 mg
Formula Weight 376.0758 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.