missing translation for 'onlineSavingsMsg'
Learn More

(2R,3R)-2-Ethyl-3-(hydroxymethyl)-4-(1-methyl-1H-imidazol-5-yl)butanoic Acid Sodium Salt (Isopilocarpic Acid Sodium Salt), Mikromol™

Product Code. 18013300
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 18013300

Brand: Mikromol MM0338.030025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(2R,3R)-2-Ethyl-3-(hydroxymethyl)-4-(1-methyl-1H-imidazol-5-yl)butanoic Acid Sodium Salt (Isopilocarpic Acid Sodium Salt)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2R,3R)-2-Ethyl-3-(hydroxymethyl)-4-(1-methyl-1H-imidazol-5-yl)butanoic Acid Sodium Salt (Isopilocarpic Acid Sodium Salt)
CAS 101769-87-3
Analyte or Component Names (2R,3R)-2-Ethyl-3-(hydroxymethyl)-4-(1-methyl-1H-imidazol-5-yl)butanoic Acid Sodium Salt (Isopilocarpic Acid Sodium Salt)
Shipping Condition Room Temperature
Molecular Formula C11 H17 N2 O3 . Na
Certifications/Compliance ISO 9001
Synonym Pilocarpine Hydrochloride Imp. C (EP), Pilocarpine Imp. C (EP), 1H-Imidazole-5-butanoic acid, alpha-ethyl-beta-(hydroxymethyl)-1-methyl-, monosodium salt, [R-(R*,R*)]- (9CI), (2R,3R)-2-Ethyl-3-(hydroxymethyl)-4-(1-methyl-1H-imidazol-5-yl)butanoic Acid Sodium Salt, Pilocarpine Hydrochloride Imp. C (EP) as Sodium Salt, Isopilocarpic Acid Sodium Salt, Pilocarpine Hydrochloride Impurity C as Sodium Salt, Pilocarpine Nitrate Impurity C as Sodium Salt, Pilocarpine Impurity C as Sodium Salt, Pilocarpine Impurity C as Sodium Salt
InChI Formula InChI=1S/C11H18N2O3.Na/c1-3-10(11(15)16)8(6-14)4-9-5-12-7-13(9)2;/h5,7-8,10,14H,3-4,6H2,1-2H3,(H,15,16);/q;+1/p-1/t8-,10+;/m0./s1
SMILES [Na+].CC[C@H]([C@H](CO)Cc1cncn1C)C(=O)[O-]
IUPAC Name sodium;(2R,3R)-2-ethyl-3-(hydroxymethyl)-4-(3-methylimidazol-4-yl)butanoate
Molecular Weight (g/mol) 248.25
Quantity 25 mg
Formula Weight 248.1137 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.