missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2RS)-1-[4-((2-Hydroxyethoxy)methyl)phenoxy]-3-(isopropylamino)-2-propanol, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
(2RS)-1-[4-((2-Hydroxyethoxy)methyl)phenoxy]-3-(isopropylamino)-2-propanol
Specifications
Specifications
| Chemical Name or Material | (2RS)-1-[4-((2-Hydroxyethoxy)methyl)phenoxy]-3-(isopropylamino)-2-propanol |
| CAS | 109791-18-6 |
| Analyte or Component Names | (2RS)-1-[4-((2-Hydroxyethoxy)methyl)phenoxy]-3-(isopropylamino)-2-propanol |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17025 |
| Synonym | (2RS)-1-[4-((2-Hydroxyethoxy)methyl)phenoxy]-3-(isopropylamino)-2-propanol |
| InChI Formula | InChI=1S/C15H25NO4/c1-12(2)16-9-14(18)11-20-15-5-3-13(4-6-15)10-19-8-7-17/h3-6,12,14,16-18H,7-11H2,1-2H3 |
| SMILES | CC(C)NCC(O)COc1ccc(COCCO)cc1 |
| IUPAC Name | 1-[4-(2-hydroxyethoxymethyl)phenoxy]-3-(propan-2-ylamino)propan-2-ol |
| Molecular Weight (g/mol) | 283.3633 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction