missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2RS)-2-Cyclohexyl-2-(4-methoxyphenyl)-N,N-dimethylethanamine Hydrochloride, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
(2RS)-2-Cyclohexyl-2-(4-methoxyphenyl)-N,N-dimethylethanamine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | (2RS)-2-Cyclohexyl-2-(4-methoxyphenyl)-N,N-dimethylethanamine Hydrochloride |
| CAS | 2108968-20-1 |
| Analyte or Component Names | (2RS)-2-Cyclohexyl-2-(4-methoxyphenyl)-N,N-dimethylethanamine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H27 N O . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Venlafaxine Hydrochloride Imp. G (EP), Venlafaxine Imp. G (EP), (2RS)-2-Cyclohexyl-2-(4-methoxyphenyl)-N,N-dimethylethanamine Hydrochloride, Venlafaxine Hydrochloride Imp. G (EP) as Hydrochloride, Venlafaxine Hydrochloride Impurity G as Hydrochloride, Venlafaxine Impurity G as Hydrochloride |
| InChI Formula | InChI=1S/C19H20F3NO2.ClH/c1-14(13-16-9-5-6-10-17(16)19(20,21)22)23-11-12-25-18(24)15-7-3-2-4-8-15;/h2-10,14,23H,11-13H2,1H3;1H |
| SMILES | Cl.COc1ccc(cc1)C(CN(C)C)C2CCCCC2 |
| IUPAC Name | 2-cyclohexyl-2-(4-methoxyphenyl)-N,N-dimethylethanamine;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction