missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(2-Methyl-5-nitro-1H-imidazol-1-yl)-1,2-epoxypropane, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
3-(2-Methyl-5-nitro-1H-imidazol-1-yl)-1,2-epoxypropane
Specifications
Specifications
| Chemical Name or Material | 3-(2-Methyl-5-nitro-1H-imidazol-1-yl)-1,2-epoxypropane |
| CAS | 16773-52-7 |
| Analyte or Component Names | 3-(2-Methyl-5-nitro-1H-imidazol-1-yl)-1,2-epoxypropane |
| Shipping Condition | Room Temperature |
| Molecular Formula | C7 H9 N3 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 3-(2-Methyl-5-nitro-1H-imidazol-1-yl)-1,2-epoxypropane |
| InChI Formula | InChI=1S/C7H9N3O3/c1-5-8-2-7(10(11)12)9(5)3-6-4-13-6/h2,6H,3-4H2,1H3 |
| SMILES | Cc1ncc([N+](=O)[O-])n1CC2CO2 |
| IUPAC Name | 2-methyl-5-nitro-1-(oxiran-2-ylmethyl)imidazole |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction