missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(4,7-Dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxybenzenesulfonic Acid, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
3-(4,7-Dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxybenzenesulfonic Acid
Specifications
Specifications
| Chemical Name or Material | 3-(4,7-Dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxybenzenesulfonic Acid |
| CAS | 1357931-55-5 |
| Analyte or Component Names | 3-(4,7-Dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxybenzenesulfonic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H20 N4 O5 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | 4-Ethoxy-3-(1-methyl-7-oxo-3-propyl-6,7-dihydro-1Hpyrazolo[4,3-d]pyrimidin-5-yl)benzenesulfonic Acid, Sildenafil Citrate Imp. D (Pharmeuropa), 3-(4,7-Dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxybenzenesulfonic Acid, Sildenafil Citrate Impurity D, Sildenafil Impurity D |
| InChI Formula | InChI=1S/C17H20N4O5S/c1-4-6-12-14-15(21(3)20-12)17(22)19-16(18-14)11-9-10(27(23,24)25)7-8-13(11)26-5-2/h7-9H,4-6H2,1-3H3,(H,18,19,22)(H,23,24,25) |
| SMILES | CCCc1nn(C)c2C(=O)NC(=Nc12)c3cc(ccc3OCC)S(=O)(=O)O |
| IUPAC Name | 4-ethoxy-3-(1-methyl-7-oxo-3-propyl-6H-pyrazolo[4,3-d]pyrimidin-5-yl)benzenesulfonic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction