missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-O-Demethylthiocolchicine, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
3-O-Demethylthiocolchicine
Specifications
Specifications
| Chemical Name or Material | 3-Demethyl Thiocolchicine |
| CAS | 87424-25-7 |
| Analyte or Component Names | 3-Demethyl Thiocolchicine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H23 N O5 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Acetamide, N-[(7S)-5,6,7,9-tetrahydro-3-hydroxy-1,2-dimethoxy-10-(methylthio)-9-oxobenzo[a]heptalen-7-yl]-, Acetamide, N-[5,6,7,9-tetrahydro-3-hydroxy-1,2-dimethoxy-10-(methylthio)-9-oxobenzo[a]heptalen-7-yl]-, (S)-, Colchicine, 2-demethylthio- (6CI), Benzo[a]heptalene, acetamide deriv., N-[(7S)-5,6,7,9-Tetrahydro-3-hydroxy-1,2-dimethoxy-10-(methylthio)-9-oxobenzo[a]heptalen-7-yl]acetamide, 3-Demethylthiocolchicine, 3-Desmethylthiocolchicine, 3-O-Demethylthiocolchicine, NSC 361792, Thiocolchicoside Impurity B |
| InChI Formula | InChI=1S/C21H23NO5S/c1-11(23)22-15-7-5-12-9-17(25)20(26-2)21(27-3)19(12)13-6-8-18(28-4)16(24)10-14(13)15/h6,8-10,15,25H,5,7H2,1-4H3,(H,22,23)/t15-/m0/s1 |
| SMILES | COc1c(O)cc2CC[C@H](NC(=O)C)C3=CC(=O)C(=CC=C3c2c1OC)SC |
| IUPAC Name | N-[(7S)-3-hydroxy-1,2-dimethoxy-10-methylsulfanyl-9-oxo-6,7-dihydro-5H-benzo[a]heptalen-7-yl]acetamide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction