missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Phenylpyrazin-2-ol, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
3-Phenylpyrazin-2-ol
Specifications
Specifications
| Chemical Name or Material | 3-Phenylpyrazin-2-ol |
| CAS | 73200-73-4 |
| Analyte or Component Names | 3-Phenylpyrazin-2-ol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H8 N2 O |
| Certifications/Compliance | ISO 17025 |
| Synonym | Ampicillin Sodium Imp. H (EP), Ampicillin Imp. H (EP), Cefaclor Imp. F (EP), 3-Phenylpyrazin-2-ol, Ampicillin Sodium Impurity H, Cefaclor Impurity F, Ampicillin Impurity H |
| InChI Formula | InChI=1S/C10H8N2O/c13-10-9(11-6-7-12-10)8-4-2-1-3-5-8/h1-7H,(H,12,13) |
| SMILES | Oc1nccnc1c2ccccc2 |
| IUPAC Name | 3-phenylpyrazin-2-ol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction