missing translation for 'onlineSavingsMsg'
Learn More

(3R)-N-Methyl-3-(naphthalen-1-yloxy)-3-(thiophen-2-yl)propan-1-amine Hydrochloride ((R)-Duloxetine Hydrochloride), Mikromol™

Product Code. 17920356
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 17920356

Brand: Mikromol MM1122.110025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(3R)-N-Methyl-3-(naphthalen-1-yloxy)-3-(thiophen-2-yl)propan-1-amine Hydrochloride ((R)-Duloxetine Hydrochloride)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (R)-Duloxetine Hydrochloride
CAS 910138-96-4
Analyte or Component Names (R)-Duloxetine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C18 H19 N O S . Cl H
Certifications/Compliance ISO 17025
Synonym (R)-Duloxetine Hydrochloride, (R)-N-Methyl-3-(naphthalen-1-yloxy)-3-(thiophen-2-yl)propan-1-amine Hydrochloride, Duloxetine Hydrochloride Imp. A (EP) as Hydrochloride, (3R)-N-Methyl-3-(naphthalen-1-yloxy)-3-(thiophen-2-yl)propan-1-amine Hydrochloride, Duloxetine Hydrochloride Imp. A (EP), Duloxetine Imp. A (EP), Duloxetine USP Related Compound A, Duloxetine USP RC A, Duloxetine Related Compound A as Hydrochloride, Duloxetine Hydrochloride Impurity A as Hydrochloride, Duloxetine Impurity A as Hydrochloride
InChI Formula InChI=1S/C18H19NOS.ClH/c1-19-12-11-17(18-10-5-13-21-18)20-16-9-4-7-14-6-2-3-8-15(14)16;/h2-10,13,17,19H,11-12H2,1H3;1H/t17-;/m1./s1
SMILES Cl.CNCC[C@@H](Oc1cccc2ccccc12)c3cccs3
IUPAC Name (3R)-N-methyl-3-naphthalen-1-yloxy-3-thiophen-2-ylpropan-1-amine;hydrochloride
Molecular Weight (g/mol) 333.88
Quantity 25 mg
Formula Weight 333.0954 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.