missing translation for 'onlineSavingsMsg'
Learn More

(3RS,4SR)-3-[(1,3-Benzodioxol-5-yloxy)methyl]-4-phenylpiperidine Hydrochloride (rac-trans-Desfluoroparoxetine Hydrochloride), Mikromol™

Product Code. 30049449
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 30049449

Brand: Mikromol MM1032.190025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(3RS,4SR)-3-[(1,3-Benzodioxol-5-yloxy)methyl]-4-phenylpiperidine Hydrochloride (rac-trans-Desfluoroparoxetine Hydrochloride)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac-trans-Desfluoroparoxetine Hydrochloride
Analyte or Component Names rac-trans-Desfluoroparoxetine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C19 H21 N O3 . Cl H
Certifications/Compliance ISO 17025
Synonym Paroxetine Related Compound B, (3RS,4SR)-3-[(1,3-Benzodioxol-5-yloxy)methyl]-4-phenylpiperidine Hydrochloride, Paroxetine Related Compound B as Hydrochloride, Paroxetine Hydrochloride Impurity A as racemic Hydrochloride, Paroxetine Impurity A as racemic Hydrochloride
InChI Formula InChI=1S/C19H21NO3.ClH/c1-2-4-14(5-3-1)17-8-9-20-11-15(17)12-21-16-6-7-18-19(10-16)23-13-22-18;/h1-7,10,15,17,20H,8-9,11-13H2;1H/t15-,17-;/m0./s1
SMILES Cl.C(Oc1ccc2OCOc2c1)[C@@H]3CNCC[C@H]3c4ccccc4
IUPAC Name (3S,4R)-3-(1,3-benzodioxol-5-yloxymethyl)-4-phenylpiperidine;hydrochloride
Molecular Weight (g/mol) 347.84
Quantity 25 mg
Formula Weight 347.1288 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.