missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-N-(2-hydroxyethyl)benzamide, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
4-Chloro-N-(2-hydroxyethyl)benzamide
Specifications
Specifications
| Chemical Name or Material | 4-Chloro-N-(2-hydroxyethyl)benzamide |
| CAS | 7400-54-6 |
| Analyte or Component Names | 4-Chloro-N-(2-hydroxyethyl)benzamide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H10 Cl N O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 4-Chloro-N-(2-hydroxyethyl)-benzamide, Benzamide, p-chloro-N-(2-hydroxyethyl)- (6CI,7CI), 2-(4-Chlorobenzoylamino)ethanol, 4-Chloro-N-(2-hydroxyethyl)benzamide, NSC 33145, Ro 18-1675 |
| InChI Formula | InChI=1S/C9H10ClNO2/c10-8-3-1-7(2-4-8)9(13)11-5-6-12/h1-4,12H,5-6H2,(H,11,13) |
| SMILES | OCCNC(=O)c1ccc(Cl)cc1 |
| IUPAC Name | 4-chloro-N-(2-hydroxyethyl)benzamide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction