missing translation for 'onlineSavingsMsg'
Learn More

4-(Dimethylamino)-1-[(1RS)-1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydroisobenzofuran-5-yl]butan-1-one Dihydrobromide, Mikromol™

Product Code. 17932706
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 17932706

Brand: Mikromol MM0242.280025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-(Dimethylamino)-1-[(1RS)-1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydroisobenzofuran-5-yl]butan-1-one Dihydrobromide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(Dimethylamino)-1-[(1RS)-1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydroisobenzofuran-5-yl]butan-1-one Dihydrobromide
CAS 2575516-42-4
Analyte or Component Names 4-(Dimethylamino)-1-[(1RS)-1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydroisobenzofuran-5-yl]butan-1-one Dihydrobromide
Shipping Condition Room Temperature
Molecular Formula C25 H33 F N2 O2 . 2 Br H
Certifications/Compliance ISO 17025
Synonym 4-(Dimethylamino)-1-[(1RS)-1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydroisobenzofuran-5-yl]butan-1-one Dihydrobromide, Citalopram Hydrobromide Impurity G as Dihydrobromide, Escitalopram Oxalate Impurity G as Dihydrobromide, Citalopram Impurity G as Dihydrobromide, Escitalopram Impurity G as Dihydrobromide
InChI Formula InChI=1S/C25H33FN2O2.2BrH/c1-27(2)15-5-7-24(29)19-8-13-23-20(17-19)18-30-25(23,14-6-16-28(3)4)21-9-11-22(26)12-10-21;;/h8-13,17H,5-7,14-16,18H2,1-4H3;2*1H
SMILES Br.Br.CN(C)CCCC(=O)c1ccc2c(COC2(CCCN(C)C)c3ccc(F)cc3)c1
IUPAC Name 4-(dimethylamino)-1-[1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-3H-2-benzofuran-5-yl]butan-1-one;dihydrobromide
Molecular Weight (g/mol) 574.36
Quantity 25 mg
Formula Weight 572.1049 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.