missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
4-Hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide
Specifications
Specifications
| Chemical Name or Material | 4-Hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide |
| CAS | 24683-25-8 |
| Analyte or Component Names | 4-Hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H10 N2 O4 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | 4-Hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide, Piroxicam Imp. C (EP), Piroxicam Impurity C |
| InChI Formula | InChI=1S/C10H10N2O4S/c1-12-8(10(11)14)9(13)6-4-2-3-5-7(6)17(12,15)16/h2-5,13H,1H3,(H2,11,14) |
| SMILES | O=C(N)C1=C(O)C=2C=CC=CC2S(=O)(=O)N1C |
| IUPAC Name | 4-hydroxy-2-methyl-1,1-dioxo-1λ^{6},2-benzothiazine-3-carboxamide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction