missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrosomorpholine, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
4-Nitrosomorpholine
Specifications
Specifications
| Chemical Name or Material | 4-Nitrosomorpholine |
| CAS | 59-89-2 |
| Analyte or Component Names | 4-Nitrosomorpholine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C4 H8 N2 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 4-Nitrosomorpholine, NSC 139, Nitrosomorpholine, Molsidomine Imp. B (EP), Morpholine, 4-nitroso-, N-Nitrosomorpholine, Molsidomine Impurity B |
| InChI Formula | InChI=1S/C4H8N2O2/c7-5-6-1-3-8-4-2-6/h1-4H2 |
| SMILES | O=NN1CCOCC1 |
| IUPAC Name | 4-nitrosomorpholine |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction