missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(tert-Butyl)benzenesulfonamide, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
4-(tert-Butyl)benzenesulfonamide
Specifications
Specifications
| Chemical Name or Material | 4-(tert-butyl)benzenesulfonamide |
| CAS | 6292-59-7 |
| Analyte or Component Names | 4-(tert-butyl)benzenesulfonamide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H15 N O2 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | Benzenesulfonamide, 4-(1,1-dimethylethyl)-, Benzenesulfonamide, p-tert-butyl- (6CI,8CI), 4-(1,1-Dimethylethyl)benzenesulfonamide, 4-tert-Butylbenzenesulfonamide, NSC 9911, p-(tert-Butyl)phenylsulfonamide, p-tert-Butylbenzenesulfonamide, (4-(tert-butyl)benzenesulfonamide), Bosentan Related Compound E, Bosentan Related Compound E |
| InChI Formula | InChI=1S/C10H15NO2S/c1-10(2,3)8-4-6-9(7-5-8)14(11,12)13/h4-7H,1-3H3,(H2,11,12,13) |
| SMILES | CC(C)(C)c1ccc(cc1)S(=O)(=O)N |
| IUPAC Name | 4-tert-butylbenzenesulfonamide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction