Learn More
Donepezil EP Impurity D, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Donepezil EP Impurity D
Specifications
Specifications
| Chemical Name or Material | 5,6-Dimethoxy-2-[(4-pyridyl)methyl]indan-1-one |
| CAS | 4803-57-0 |
| Analyte or Component Names | 5,6-Dimethoxy-2-[(4-pyridyl)methyl]indan-1-one |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H17 N O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 5,6-Dimethoxy-2-[(4-pyridyl)methyl]indan-1-one, 2,3-Dihydro-5,6-dimethoxy-2-(4-pyridinylmethyl)-1H-inden-1-one, 5,6-Dimethoxy-2-(4-pyridylmethyl)-1-indanone, Donepezil Hydrochloride Impurity D, Donepezil Impurity D |
| InChI Formula | InChI=1S/C17H17NO3/c1-20-15-9-12-8-13(7-11-3-5-18-6-4-11)17(19)14(12)10-16(15)21-2/h3-6,9-10,13H,7-8H2,1-2H3 |
| SMILES | COc1cc2CC(Cc3ccncc3)C(=O)c2cc1OC |
| IUPAC Name | 5,6-dimethoxy-2-(pyridin-4-ylmethyl)-2,3-dihydroinden-1-one |
| Show More |