missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlordiazepoxide EP Impurity B, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
Chlordiazepoxide EP Impurity B
Specifications
Specifications
| Chemical Name or Material | 6-Chloro-2-(chloromethyl)-4-phenylquinazoline 3-Oxide |
| CAS | 5958-24-7 |
| Analyte or Component Names | 6-Chloro-2-(chloromethyl)-4-phenylquinazoline 3-Oxide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H10 Cl2 N2 O |
| Certifications/Compliance | ISO 17025 |
| Synonym | Chlordiazepoxide Hydrochloride Imp. B (EP), Chlordiazepoxide Imp. B (EP), 6-Chloro-2-(chloromethyl)-4-phenylquinazoline 3-Oxide, Chlordiazepoxide Hydrochloride Impurity B, Chlordiazepoxide Impurity B, Chlordiazepoxide Impurity B |
| InChI Formula | InChI=1S/C15H10Cl2N2O/c16-9-14-18-13-7-6-11(17)8-12(13)15(19(14)20)10-4-2-1-3-5-10/h1-8H,9H2 |
| SMILES | [O-][n+]1c(CCl)nc2ccc(Cl)cc2c1c3ccccc3 |
| IUPAC Name | 6-chloro-2-(chloromethyl)-3-oxido-4-phenylquinazolin-3-ium |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction