missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7H-Purin-6-amine Sulphate (Adenine Sulphate), Mikromol
Click to view available options
Quantity:
25 mg
4 x 25 mg
Unit Size:
25mg
Description
7H-Purin-6-amine Sulphate (Adenine Sulphate)
Specifications
Specifications
| Chemical Name or Material | Adenine Sulphate |
| CAS | 321-30-2 |
| Analyte or Component Names | Adenine Sulphate |
| Shipping Condition | Room Temperature |
| Molecular Formula | 2[C5 H5 N5] . S O4 H2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Adenosine Imp. A (EP), 7H-Purin-6-amine Sulphate, Adenosine Imp. A (EP) as Sulphate, Adenine Sulphate, Adenosine Impurity A as Sulphate |
| InChI Formula | InChI=1S/2C5H5N5.H2O4S/c2*6-4-3-5(9-1-7-3)10-2-8-4;1-5(2,3)4/h2*1-2H,(H3,6,7,8,9,10);(H2,1,2,3,4) |
| SMILES | Nc1ncnc2[nH]cnc12.Nc3ncnc4[nH]cnc34.OS(=O)(=O)O |
| IUPAC Name | 9H-purin-6-amine;sulfuric acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction