missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aceclofenac Isopropyl Ester, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Aceclofenac Isopropyl Ester
Specifications
Specifications
| Chemical Name or Material | Aceclofenac Isopropyl Ester |
| CAS | 2514625-79-5 |
| Analyte or Component Names | Aceclofenac Isopropyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H19 Cl2 N O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Aceclofenac Isopropyl Ester |
| InChI Formula | InChI=1S/C19H19Cl2NO4/c1-12(2)26-18(24)11-25-17(23)10-13-6-3-4-9-16(13)22-19-14(20)7-5-8-15(19)21/h3-9,12,22H,10-11H2,1-2H3 |
| SMILES | CC(C)OC(=O)COC(=O)Cc1ccccc1Nc2c(Cl)cccc2Cl |
| IUPAC Name | (2-oxo-2-propan-2-yloxyethyl) 2-[2-(2,6-dichloroanilino)phenyl]acetate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction