missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetylsalicylsalicylsalicylsalicylic Acid, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Acetylsalicylsalicylsalicylsalicylic Acid
Specifications
Specifications
| Chemical Name or Material | Acetylsalicylsalicylsalicylsalicylic Acid |
| CAS | 85539-30-6 |
| Analyte or Component Names | Acetylsalicylsalicylsalicylsalicylic Acid |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C30 H20 O10 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Benzoic acid, 2-[[2-(acetyloxy)benzoyl]oxy]-, 2-[(2-carboxyphenoxy)carbonyl]phenyl ester, Acetylsalicylsalicylsalicylsalicylic Acid |
| InChI Formula | InChI=1S/C30H20O10/c1-18(31)37-24-15-7-3-11-20(24)28(34)39-26-17-9-5-13-22(26)30(36)40-25-16-8-4-12-21(25)29(35)38-23-14-6-2-10-19(23)27(32)33/h2-17H,1H3,(H,32,33) |
| SMILES | CC(=O)Oc1ccccc1C(=O)Oc2ccccc2C(=O)Oc3ccccc3C(=O)Oc4ccccc4C(=O)O |
| IUPAC Name | 2-[2-[2-(2-acetyloxybenzoyl)oxybenzoyl]oxybenzoyl]oxybenzoic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction