missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azoxystrobin-d4, TRC
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Chemical Name or Material | Azoxystrobin d4 |
| CAS | 1346606-39-0 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C22 2H4 H13 N3 O5 |
| Quantity | 25 mg |
| Synonym | Methyl 2-[2-[6-(2-cyano-3,4,5,6-tetradeuteriophenoxy)pyrimidin-4-yl]oxyphenyl]-3-methoxyprop-2-enoate |
| InChI Formula | InChI=1S/C22H17N3O5/c1-27-13-17(22(26)28-2)16-8-4-6-10-19(16)30-21-11-20(24-14-25-21)29-18-9-5-3-7-15(18)12-23/h3-11,13-14H,1-2H3/b17-13+/i3D,5D,7D,9D |
| SMILES | [2H]c1c([2H])c([2H])c(C#N)c(Oc2cc(Oc3ccccc3\C(=C/OC)\C(=O)OC)ncn2)c1[2H] |
| IUPAC Name | methyl (E)-2-[2-[6-(2-cyano-3,4,5,6-tetradeuteriophenoxy)pyrimidin-4-yl]oxyphenyl]-3-methoxyprop-2-enoate |
| Molecular Weight (g/mol) | 407.41 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction