missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 3-Fluoro-4-(4-morpholinyl)phenylcarbamate, Mikromol™

Product Code. 30050664
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 30050664

Brand: Mikromol MM3300.180025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Benzyl 3-Fluoro-4-(4-morpholinyl)phenylcarbamate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzyl 3-Fluoro-4-(4-morpholinyl)phenylcarbamate
CAS 168828-81-7
Analyte or Component Names Benzyl 3-Fluoro-4-(4-morpholinyl)phenylcarbamate
Shipping Condition Room Temperature
Molecular Formula C18 H19 F N2 O3
Certifications/Compliance ISO 17025
Synonym N-[3-Fluoro-4-(4-morpholinyl)phenyl]carbamic Acid Phenylmethyl Ester, Carbamic acid, [3-fluoro-4-(4-morpholinyl)phenyl]-, phenylmethyl ester (9CI), (3-Fluoro-4-morpholin-4-ylphenyl)carbamic acid benzyl ester, Benzyl 3-fluoro-4-(4-morpholinyl)phenylcarbamate, N-(Benzyloxycarbonyl)-3-fluoro-4-(4-morpholinyl)aniline, N-Benzyloxycarbonyl-3-fluoro-4-morpholinoaniline, [3-Fluoro-4-(4-morpholinyl)phenyl]carbamic acid phenylmethyl ester
InChI Formula InChI=1S/C18H19FN2O3/c19-16-12-15(6-7-17(16)21-8-10-23-11-9-21)20-18(22)24-13-14-4-2-1-3-5-14/h1-7,12H,8-11,13H2,(H,20,22)
SMILES Fc1cc(NC(=O)OCc2ccccc2)ccc1N3CCOCC3
IUPAC Name benzyl N-(3-fluoro-4-morpholin-4-ylphenyl)carbamate
Molecular Weight (g/mol) 330.35
Quantity 25 mg
Formula Weight 330.138 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.