missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmethyltrifluoperazine Dihydrochloride, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Desmethyltrifluoperazine Dihydrochloride
Specifications
Specifications
| Chemical Name or Material | Desmethyltrifluoperazine Dihydrochloride |
| CAS | 4617-82-7 |
| Analyte or Component Names | Desmethyltrifluoperazine Dihydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H22 F3 N3 S . 2 Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | 10H-Phenothiazine, 10-[3-(1-piperazinyl)propyl]-2-(trifluoromethyl)-, hydrochloride (1:2), Phenothiazine, 10-[3-(1-piperazinyl)propyl]-2-(trifluoromethyl)-, dihydrochloride (6CI,8CI), Desmethyltrifluoperazine Dihydrochloride |
| InChI Formula | InChI=1S/C20H22F3N3S.2ClH/c21-20(22,23)15-6-7-19-17(14-15)26(16-4-1-2-5-18(16)27-19)11-3-10-25-12-8-24-9-13-25;;/h1-2,4-7,14,24H,3,8-13H2;2*1H |
| SMILES | Cl.Cl.FC(F)(F)c1ccc2Sc3ccccc3N(CCCN4CCNCC4)c2c1 |
| IUPAC Name | 10-(3-piperazin-1-ylpropyl)-2-(trifluoromethyl)phenothiazine;dihydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu