missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(E)-3-(Dibenzo[b,e]oxepin-11(6H)-ylidene)-N-methylpropan-1-amine Hydrochloride (Desmethyldoxepin Hydrochloride), Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
(E)-3-(Dibenzo[b,e]oxepin-11(6H)-ylidene)-N-methylpropan-1-amine Hydrochloride (Desmethyldoxepin Hydrochloride)
Specifications
Specifications
| Chemical Name or Material | Doxepin Related Compound C |
| CAS | 4504-96-5 |
| Analyte or Component Names | Doxepin Related Compound C |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H19 N O . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | E-3-(dibenzo[b,e]oxepin-11(6H)-ylidene)-N-methylpropan-1-amine hydrochloride, N-Desmethyl (E)-Doxepin Hydrochloride, Doxepin Related Compound C as Hydrochloride, Doxepin Hydrochloride Impurity C as Hydrochloride, Doxepin Impurity C as Hydrochloride |
| InChI Formula | InChI=1S/C18H19NO.ClH/c1-19-12-6-10-16-15-8-3-2-7-14(15)13-20-18-11-5-4-9-17(16)18;/h2-5,7-11,19H,6,12-13H2,1H3;1H/b16-10+; |
| SMILES | Cl.CNCC\C=C\1/c2ccccc2COc3ccccc13 |
| IUPAC Name | (3E)-3-(6H-benzo[c][1]benzoxepin-11-ylidene)-N-methylpropan-1-amine;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction