missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Estradiol 3-Methyl Ether, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Estradiol 3-Methyl Ether
Specifications
Specifications
| Chemical Name or Material | Estradiol 3-Methyl Ether |
| CAS | 1035-77-4 |
| Analyte or Component Names | Estradiol 3-Methyl Ether |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H26 O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Estra-1,3,5(10)-trien-17-ol, 3-methoxy-, (17β)-, Estra-1,3,5(10)-trien-17β-ol, 3-methoxy- (6CI,8CI), (17β)-3-Methoxyestra-1,3,5(10)-trien-17-ol, 17β-Estradiol 3-methyl ether, 17β-Hydroxy-3-methoxyestra-1,3,5(10)-triene, 3-Methoxy-13β-methyl-1,3,5(10)-gonatrien-17β-ol, 3-Methoxy-17β-hydroxyestra-1,3,5(10)-triene, 3-Methoxyestra-1,3,5(10)-trien-17β-ol, 3-Methoxyestra-1,3,5(10)-triene-17β-ol, 3-Methoxyestra-1,3,5(10)-triene-3,17β-diol, 3-Methoxyestradiol, 3-Methoxyoestra-1,3,5(10)-trien-17β-ol, 3-O-Methylestradiol, Estradiol 3-methyl ether, NSC 58851 |
| InChI Formula | InChI=1S/C19H26O2/c1-19-10-9-15-14-6-4-13(21-2)11-12(14)3-5-16(15)17(19)7-8-18(19)20/h4,6,11,15-18,20H,3,5,7-10H2,1-2H3/t15-,16-,17+,18+,19+/m1/s1 |
| SMILES | COc1ccc2[C@H]3CC[C@]4(C)[C@@H](O)CC[C@H]4[C@@H]3CCc2c1 |
| IUPAC Name | (8R,9S,13S,14S,17S)-3-methoxy-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-17-ol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction